SpectraMax® iD3 Multi-Mode Microplate Reader, Molecular Devices | Avantor
SpectraMax® iD3 Multi-Mode Microplate Reader, Molecular Devices
SpectraMax® iD3 Multi-Mode Microplate Reader
Catalog # 75886-128
Fournisseur #:  ID3-STD
SpectraMax® iD3 Multi-Mode Microplate Reader, Molecular Devices
Catalog # 75886-128
Supplier:  
SpectraMax® iD3 Multi-Mode Microplate Reader, Molecular Devices
SpectraMax® iD3 Multi-Mode Microplate Reader, Molecular Devices
SpectraMax® iD3 Multi-Mode Microplate Reader
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
PLATFORM ID3 MULTI-MODE MICPLT DETECTION Catalog #CA75886-128
Le produit CA75886-128 est un remplacement de 75886-128 et a le même emballage et les mêmes spécifications, mais peut être d'une quantité différente.
Product Details & Documents
Unparalleled performance on a personalized platform. The SpectraMax® iD3 Multi-Mode Microplate Reader is the cornerstone of a complete laboratory solution designed to expand the boundaries of research capabilities. With optimized reagents and the industry-leading data acquisition and analysis tool, SoftMax Pro® 7 Software, the SpectraMax® iD3 allows users to customize workflow to perfectly match their needs.
  • Use the touchscreen to easily set up methods, run experiments, or view tutorial videos
  • Personalize workflows with one-tap near-field communication (NFC) functionality
  • Push data to workstations, eliminating the need to retrieve data directly from the instrument
  • Validate the instrument and software with an extensive suite of tools
The SpectraMax® iD3 Multi-Mode Microplate Reader features a large, high-resolution touchscreen with an embedded software package that enables users to customize protocols, take advantage of preconfigured protocols, and run experiments without the need for a dedicated computer workstation
View data quickly using the large touchscreen, export to a USB drive, or analyze data using the industry-leading data acquisition and analysis tool, SoftMax Pro® 7 Software. QuickSync technology allows users to walk away from the SpectraMax® iD3 to focus on additional research. Data is automatically delivered to a workstation, eliminating the need to physically retrieve data from the instrument. Featuring orbital shaking, a four-monochromator optical pathway with high efficiency gratings, well scanning up to a 20x20 read matrix, spectral scanning and detection of plate formats from 6- to 384-wells, the SpectraMax® iD3 reader is a complete research solution.
Expand any lab’s capabilities to include flash applications such as dual luciferase and ATP assays with the SpectraMax® iD3 injector system with SmartInject™ Technology ensuring equal mixing across the plate for high-precision experiments. Engineered for high performance, the dual injector system can be used with any read mode and includes safeguard features such as overflow protection to save precious reagents and maximize performance for those critical experiments in the lab. SoftMax Pro® GxP Compliance Software extends Molecular Devices leading data acquisition and analysis solution into regulated laboratories working under GMP, GLP, 21 CFR Part 11, and other similar guidelines for secure electronic records.
A built-in near-field communication (NFC) reader offers access to custom protocols with a single tap, saving precious time. Pair NFC Tags with specific user profiles to gain access to personalized protocols and experiment results, eliminating the need to scroll through dozens of files. The superior optical system includes a xenon flash lamp and features an ultra-cooled photomultiplier tube (PMT) to –5 °C that reduces background noise for excellent sensitivity and a wide dynamic range. Simple-to-use temperature control provides adjustment to experimental conditions, from ambient up to 65 °C, expanding laboratory capabilities to include temperature sensitive assays.
Certifications: Meets ASTM standards. EN-342.
Ordering information: SpectraMax® iD3 Multi-Mode Microplate Reader comes standard with Absorbance, Fluorescence, and Luminescence as well as a superior optical system that includes a xenon flash lamp.
Spécifications
  • Désignation:
    SpectraMax® iD3 Multi-Mode Microplate Reader