Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Spare sealing, 5 pieces, for rotor Lids FA-45-24-11 and FA-45-16-17
Catalog # 75875-158
Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Catalog # 75875-158
Supplier:  
Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Spare sealing, 5 pieces, for rotor Lids FA-45-24-11 and FA-45-16-17
Discontinued
Product Details & Documents
Spécifications
  • Désignation:
    Spare sealing, 5 pieces, for rotor Lids FA-45-24-11 and FA-45-16-17
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Rotor key, 5 mm
    Rotor key, 5 mm
    CA53513-248
    Centrifuges 5417 C/R, 5418/R, 5424/R, 5430/R and 5702/R/RH
    Log in pour la disponibilité
    Each - $42.30