Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Basic Industrial Coveralls

VWR® Basic Industrial Coveralls

Green, M, 67" (body), 29" (leg), 26"
Catalog # 75874-268
Supplier:   VWR International
VWR® Basic Industrial Coveralls
Catalog # 75874-268
Supplier:  VWR International
VWR® Basic Industrial Coveralls
VWR® Basic Industrial Coveralls

Green, M, 67" (body), 29" (leg), 26"

Price
$125.18
New Customer Promotional Price
Log in for availability
Case of 25
Product Details & Documents

Disposable coverall made from 20 gram spunbond polypropylene (SPP) fabric. Provides lightweight, breathable protection for nonhazardous environments. Ideal for general industrial use, food processing, and clean areas requiring basic contamination control.

  • Lightweight and breathable fabric reduces heat buildup during extended use

  • Zipper front closure for easy donning and removal
  • Elastic wrists and ankles for better coverage
  • Single-use, disposable garment for contamination control
  • Economical solution for basic protection needs
  • Non-linting material helps maintain clean environments
  • Suitable for general industrial, food processing, and nonhazardous environments

The VWR® basic protection coverall is designed to provide reliable, economical protection in environments where contamination control is essential, but exposure risks are minimal. Constructed from 20 gram spunbond polypropylene (SPP), this coverall offers a lightweight and breathable solution that ensures user comfort during extended wear. The non-woven SPP fabric is non-linting, reducing the risk of particle contamination, and is suitable for clean areas, food processing, and general industrial applications.

Key design features include a zipper front closure for easy donning and removal, along with elastic wrists and ankles to enhance coverage and minimize gaps. The disposable nature of the garment supports hygiene compliance.

Packaging: Coveralls are packaged 25 per case.

Documents

Specifications
  • Taille de confection:
    M
  • Color:
    Green
  • Longueur:
    67" (body), 29" (leg)
  • Chest Size:
    26"
  • Cat. No.:
    75874-268
  • Cleanroom class:
    10000/ISO 7
  • Sleeve or leg length:
    Long
  • Garment application:
    Cleanroom|Industrial|Laboratory
  • Gender:
    Unisex
  • Pockets:
    Without pockets
  • Closure type:
    Zip closure
  • Fabric:
    Spunbound polypropylene
  • Particle shedding (Helmke Drum):
    Level II
  • Water resistance:
    First line barrier
  • Weight:
    20 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.