Section Block, Electron Microscopy Sciences
Section Block, 500 mL
  103306-942
Section Block, Electron Microscopy Sciences
  103306-942
 :  
Section Block, Electron Microscopy Sciences
Section Block, 500 mL
 :  
$2,828.21
Each (500ml)
Blocking solutions based on chemically modified and fragmented ultra-pure casein. Effectively reduces unwanted binding of primary antibody and conjugates use to charged surface of the slide and tissue section.
Greatly reduces non-specific binding while preserving the specific reaction, by saturating potential non-specific protein-protein interactions.
Recommended for research and diagnostic pathology, especially for retrieved sections and polyclonal antibodies.
  • Désignation:
    Section Block
  • Taille:
    500 mL