Freeze Substitution Kit, Electron Microscopy Sciences | Avantor
Freeze Substitution Kit, Electron Microscopy Sciences
Freeze substitution kit without shaker
Catalog # 103306-432
Fournisseur #:  34500-S
Freeze Substitution Kit, Electron Microscopy Sciences
Catalog # 103306-432
Freeze Substitution Kit, Electron Microscopy Sciences
Freeze substitution kit without shaker
Price
$3,404.82
Log in pour la disponibilité
Each Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For dehydration and fixation of rapidly frozen cells.
  • Excellent freeze substitution results in as little as 90 minutes
  • Extend the time for larger samples to 3 hours by putting the lid on
Freeze Substitution is a process for low temperature dehydration and fixation of rapidly frozen cells that usually takes days to complete.
With this unique kit researchers are now able to achieve excellent freeze substitution results in as little as 90 minutes for cells of small volume such as bacteria and tissue culture cells. For those cells of greater volume or that have significant diffusion barriers such as cuticles or thick cell walls, one can extend the time to 3 hours simply by putting a lid on the box.
Kit features platform shaker, ice chest, heater block, temperature probe, cryo tubes, and data logger.
Spécifications
  • Désignation:
    Freeze substitution kit without shaker
  • Cat. No.:
    103306-432
  • Model:
    34500-S