G57 Pharmaceutical PSA Patterns, Electron Microscopy Sciences
G57 Pharmaceutical PSA Patterns, Electron Microscopy Sciences
Graticle G-57, 20.4 mm
Catalog # 103306-218
Supplier #:  68091-20.4
G57 Pharmaceutical PSA Patterns, Electron Microscopy Sciences
Catalog # 103306-218
G57 Pharmaceutical PSA Patterns, Electron Microscopy Sciences
G57 Pharmaceutical PSA Patterns, Electron Microscopy Sciences
Graticle G-57, 20.4 mm
Price
$880.01
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This graticule was designed for the pharmaceutical industry.
  • Scale is incorporated
  • Glass sandwich surface
However, it is also useful where particle size considerations are restricted to 10 and 25 mm. Dots and circles give quick references for these two sizes.
The microscope must be calibrated when ordering this graticule, such that the circle must equate to one mm on the microscope stage.
Specifications
  • Désignation:
    Graticle G-57
  • Taille:
    20.4 mm