Orbital Specimen Holder, Electron Microscopy Sciences
Product Details & Documents
Unique orbital specimen holder for a spin and raise SEM images to a whole new level.
- Rotate 360° around the axis perpendicular to the electron beam
- Position the sample with turn of a screw
- Flexibile and superior control
The pot is coated to reduce noise, producing a black background behind the subject so every detail stands out, allowing the capture of stunning, clear images.
Specifications
- Désignation:Orbital Specimen Holder M4 Mount