Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Set of Five, 0.25 – 1.25 mm
  103305-792
 :  57401
Brain Punch Kit, Electron Microscopy Sciences
  103305-792
 :  
Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Set of Five, 0.25 – 1.25 mm
The Brain Punch Kit is a set of electropolished and sharpened punches for the Palkovit's punch technique.
Each punch has an electro polished sharp end for smooth cuts and minimal tissue adhesion. The smaller tubes are reinforced by an outer layer of concentric tubing except at the tip.
  • Désignation:
    Brain Punch Set of Five, 0.25 – 1.25 mm