Section Block, Electron Microscopy Sciences | Avantor
Section Block, Electron Microscopy Sciences
Section Block, 50 mL
Catalog # 103305-762
Fournisseur #:  62710
Section Block, Electron Microscopy Sciences
Catalog # 103305-762
CAS Number:  
Section Block, Electron Microscopy Sciences
Section Block, 50 mL
CAS Number:  
Price
$361.16
Log in pour la disponibilité
Each (50ml) Kit of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Blocking solutions based on chemically modified and fragmented ultra-pure casein. Effectively reduces unwanted binding of primary antibody and conjugates use to charged surface of the slide and tissue section.
Greatly reduces non-specific binding while preserving the specific reaction, by saturating potential non-specific protein-protein interactions.
Recommended for research and diagnostic pathology, especially for retrieved sections and polyclonal antibodies.
Spécifications
Spécifications
  • Désignation:
    Section Block
  • Taille:
    50 mL