SEM Bulk Specimen Holder, Electron Microscopy Sciences
Product Details & Documents
For clamping large irregular, bulky samples to view in a scanning electron microscope.
- Made of machined aluminum with stainless steel allen set screws
- Pin Diameter: 3.2 mm
: Allen wrench included.
Spécifications
- Désignation:X-Large Bulk Specimen Holder
- Dimensions:40 x 52 x 13 mm
- Cat. No.:103305-562