Saws, Electron Microscopy Sciences | Avantor
Saws, Electron Microscopy Sciences
Charriere Saw, 267 mm Long
Catalog # 103305-476
Fournisseur #:  70566-30
Saws, Electron Microscopy Sciences
Catalog # 103305-476
Saws, Electron Microscopy Sciences
Saws, Electron Microscopy Sciences
Charriere Saw, 267 mm Long
Price
266,52 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Three styles of manual bone saws are available.
  • Broad bladed Satterlee Saw
  • Thinner rounded top Charriere Saw
  • Solid handle Langenbech Metacarpal Saw
The broad bladed Satterlee Saw (A), the thinner rounded tip Charriere Saw (B), and the solid handle Langenbech Metacarpal Saw (C). Each saw has serrated teeth.
Spécifications
  • Désignation:
    Charriere Saw, 267 mm Long