Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Individual, 1.50 mm
  103305-470
Brain Punch Kit, Electron Microscopy Sciences
  103305-470
Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Individual, 1.50 mm
$435.36
Each
The Brain Punch Kit is a set of electropolished and sharpened punches for the Palkovit's punch technique.
Each punch has an electro polished sharp end for smooth cuts and minimal tissue adhesion. The smaller tubes are reinforced by an outer layer of concentric tubing except at the tip.
  • Désignation:
    Brain Punch Individual, 1.50 mm