Brain Punch Kit, Electron Microscopy Sciences | Avantor
Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Individual, 0.50 mm
Catalog # 103305-074
Fournisseur #:  57393
Brain Punch Kit, Electron Microscopy Sciences
Catalog # 103305-074
Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Individual, 0.50 mm
Price
$448.69
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Brain Punch Kit is a set of electropolished and sharpened punches for the Palkovit's punch technique.
Each punch has an electro polished sharp end for smooth cuts and minimal tissue adhesion. The smaller tubes are reinforced by an outer layer of concentric tubing except at the tip.
Spécifications
  • Désignation:
    Brain Punch Individual, 0.50 mm