Slide Rack, Electron Microscopy Sciences
Rack Carrier, 20 Slides
  103302-682
Marketsource
 :  71435-DAKO
Slide Rack, Electron Microscopy Sciences
  103302-682
Marketsource
Slide Rack, Electron Microscopy Sciences
Slide Rack, Electron Microscopy Sciences
Rack Carrier, 20 Slides
$155.87
Each
Poly-ceramic slide rack designed for histology,cytology staining and coverslipping.
  • Made from Poly-Ceramic
  • Microwave safely in a distortion-free rack
  • Chemical resistant to all solvents below 200 °C
  • Designed – for histology and cytology staining and coverslipping
  • Tested for performance criteria and dimensional accuracy
  • Fit the Tissue-Tek Tape Machines
  • Easy slide insertion in V-Groove slots
Stainless Steel and Plastic Slide Racks are very good chemically as well as thermally but have limited uses in microwave and solvents. When it comes to inserting the slide, easy to break the glass or tear the tape on abrupt edges.
WINLAB Slide Racks are made from Poly-ceramic that maintains its structural integrity to 200 °C in any solvent while being microwaved. The racks last a lot longer than any other materials. WINLAB Racks save time and money.
  • Capacité:
    20 Slides
  • Désignation:
    Rack Carrier