Slide Rack, Electron Microscopy Sciences | Avantor
Slide Rack, Electron Microscopy Sciences
Rack Carrier, 30 slides
Catalog # 103302-680
Marketsource
Fournisseur #:  71433-DAKO
Slide Rack, Electron Microscopy Sciences
Catalog # 103302-680
Marketsource
Slide Rack, Electron Microscopy Sciences
Slide Rack, Electron Microscopy Sciences
Rack Carrier, 30 slides
Price
$160.29
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Poly-ceramic slide rack designed for histology,cytology staining and coverslipping.
  • Made from Poly-Ceramic
  • Microwave safely in a distortion-free rack
  • Chemical resistant to all solvents below 200 °C
  • Designed – for histology and cytology staining and coverslipping
  • Tested for performance criteria and dimensional accuracy
  • Fit the Tissue-Tek Tape Machines
  • Easy slide insertion in V-Groove slots
Stainless Steel and Plastic Slide Racks are very good chemically as well as thermally but have limited uses in microwave and solvents. When it comes to inserting the slide, easy to break the glass or tear the tape on abrupt edges.
WINLAB Slide Racks are made from Poly-ceramic that maintains its structural integrity to 200 °C in any solvent while being microwaved. The racks last a lot longer than any other materials. WINLAB Racks save time and money.
Spécifications
  • Capacité:
    30 slides
  • Désignation:
    Rack Carrier