Stainless Steel Baskets with Multiple Compartments, Electron Microscopy Sciences
Product Details & Documents
These stainless steel baskets with lids and multiple compartments are used for critical point drying.
- Single or multiple compartments
These baskets are either 15 or 23 mm in diameter.
Spécifications
- Désignation:4 Compartment Tissue Basket with Lid
- I.D.×H:15 mm×10 mm