Scratch Brushes with Metal Ferrules, Electron Microscopy Sciences | Avantor
Scratch Brushes with Metal Ferrules, Electron Microscopy Sciences
Fiberglass Ferrules, Fiberglass, 1/8", 4 1/4"
Catalog # 103302-490
Fournisseur #:  65582-05
Scratch Brushes with Metal Ferrules, Electron Microscopy Sciences
Catalog # 103302-490
Scratch Brushes with Metal Ferrules, Electron Microscopy Sciences
Scratch Brushes with Metal Ferrules, Electron Microscopy Sciences
Fiberglass Ferrules, Fiberglass, 1/8", 4 1/4"
Price
$41.89
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These scratch brushes with pen shaped handles are used for fine deburring, polishing, as well as rust removal and the cleaning and removal of deposited wax.
  • Available with steel, nylon, fiberglass or brass
  • With metal ferrules for improved durability
  • Overall length is 41/4" and the brush is 1/8" wide
Packaging: Refills are available and are packaged 12/box.
Spécifications
  • Bristle Material:
    Fiberglass
  • Désignation:
    Fiberglass Ferrules
  • Longueur totale:
    4 1/4"
  • Brush Head Width:
    1/8"
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Brush Refill, Nylon Ferrules
    Brush Refill, Nylon Ferrules
    103302-466
    Log in pour la disponibilité
    Each - $23.17