Multi-Well Embedding Mold, Electron Microscopy Sciences | Avantor
Multi-Well Embedding Mold, Electron Microscopy Sciences
Multi-Well Embedding Mold, Clear
Catalog # 103302-476
Marketsource
Fournisseur #:  70912
Multi-Well Embedding Mold, Electron Microscopy Sciences
Catalog # 103302-476
Marketsource
Multi-Well Embedding Mold, Electron Microscopy Sciences
Multi-Well Embedding Mold, Electron Microscopy Sciences
Multi-Well Embedding Mold, Clear
Price
$31.77
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Clear, white or blue silicone mold for embedding.
This unique silicone mold has 2 different cavity sizes to meet all embedding needs. 2 troughs: 1 L x 1/8 W x 1/8" D. 14 cavities: 1/4 L x 1/2 W x 1/8" D. 1 trough: 23/4 L x 3/32 W x 1/8" D. 10 cavities: 3/8 L x 3/4 W x 1/8" D. Each cavity has an alpha-numeric marking, A1-A7, B1-B7, C1-C5, D1-D5. Overall mold measures: 43/4 L x 3 W x 1/4" H (121 x 76 x 6 mm).
Spécifications
  • Désignation:
    Multi-Well Embedding Mold, Clear