Membrane Boxes, Electron Microscopy Sciences
Membrane box, rectangular, 175×100×75 mm, 129×54×40 mm
  103302-406
Marketsource
 :  64304-58
Membrane Boxes, Electron Microscopy Sciences
  103302-406
Marketsource
Membrane Boxes, Electron Microscopy Sciences
Membrane Boxes, Electron Microscopy Sciences
Membrane box, rectangular, 175×100×75 mm, 129×54×40 mm
$94.50
Each
These unique membrane boxes are for the transfer, storage and shipping of many delicate items. Once your sensitive item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
  • Protects items from any movement
  • Double elastic membrane on the top and bottom offers security
  • Available in round, rectangular and square shapes
  • Désignation:
    Membrane box, rectangular
  • Dimensions:
    175×100×75 mm
  • Interior Dimensions:
    129×54×40 mm