Membrane Boxes, Electron Microscopy Sciences
Membrane box, square, 125×125×75 mm, 92×92×60 mm
Catalog # 103302-386
Marketsource
 :  64304-18
Membrane Boxes, Electron Microscopy Sciences
Catalog # 103302-386
Marketsource
Membrane Boxes, Electron Microscopy Sciences
Membrane Boxes, Electron Microscopy Sciences
Membrane box, square, 125×125×75 mm, 92×92×60 mm
Price
$107.72
Log in for availability
Each Pack of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These unique membrane boxes are for the transfer, storage and shipping of many delicate items. Once your sensitive item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
  • Protects items from any movement
  • Double elastic membrane on the top and bottom offers security
  • Available in round, rectangular and square shapes
Specifications
  • Désignation:
    Membrane box, square
  • Dimensions:
    125×125×75 mm
  • Interior Dimensions:
    92×92×60 mm