SEM Clip Specimen Mounts, Electron Microscopy Sciences
SEM Clip, 25 mm Pin Mount, 1 clip, 25 mm
Catalog # 103302-342
 :  75923-12
SEM Clip Specimen Mounts, Electron Microscopy Sciences
Catalog # 103302-342
SEM Clip Specimen Mounts, Electron Microscopy Sciences
SEM Clip Specimen Mounts, Electron Microscopy Sciences
SEM Clip, 25 mm Pin Mount, 1 clip, 25 mm
Price
$69.29
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specially designed for easy, quick and clean mounting of any type of thin specimen on specimen stubs.
  • Available with single and multiple clips
  • Ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal, and more
  • SEM clip mounts with multiple clips can hold larger samples or multiple smaller samples
These spring-loaded clips accommodate samples of up to 2 mm thickness, samples are held securely and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time.
Spécifications
  • Ø:
    25 mm
  • Désignation:
    SEM Clip, 25 mm Pin Mount, 1 clip