Earth-Friendly Long Handle Sampling Spoons, Electron Microscopy Sciences
Product Details & Documents
Long handles ideal for acquiring samples from bottles, jars, and more.
- Length 17.8 cm
- Non-sterile
- Translucent
- Available in 3 bowl sizes
Long-handled spoons are ideal for acquiring samples from bottles, jars, and other confined areas and helping to avoid personal contact with sample materials. A flat edge on the rim of each bowl allows fast levelling to collect and measure small amounts with accuracy. Each spoon is 17.8 cm (7") overall length. Non-sterile and translucent in color.
Spécifications
- Capacité:½ tsp.
- Désignation:Sample Spoon