Disposable Multi-Well Embedding Mold, Electron Microscopy Sciences
Product Details & Documents
Disposable flexible LDPE mold for embedding.
A clear flexible LDPE mold to clean the molds. 2 troughs: 1 L x 1/8 W x 1/8" D. 14 cavities: 1/4 L x 1/2 W x 1/8" D. 1 trough: 23/4 L x 3/32 W x 1/8 " D. 10 cavities: 3/8 L x 3/4 W x 1/8" D.
Spécifications
- Désignation:Disposable Multi-Well Embedding Mold