Saws, Electron Microscopy Sciences
Product Details & Documents
Three styles of manual bone saws are available.
- Broad bladed Satterlee Saw
- Thinner rounded top Charriere Saw
- Solid handle Langenbech Metacarpal Saw
The broad bladed Satterlee Saw (A), the thinner rounded tip Charriere Saw (B), and the solid handle Langenbech Metacarpal Saw (C). Each saw has serrated teeth.
Specifications
- Désignation:Satterlee Saw Replacement Blade