Athene Square Grids, Electron Microscopy Sciences | Avantor
Athene Square Grids, Electron Microscopy Sciences
Square Grids, Nickel, 3.05 mm, 300
Catalog # 103302-122
Fournisseur #:  A300-NI
Athene Square Grids, Electron Microscopy Sciences
Catalog # 103302-122
Athene Square Grids, Electron Microscopy Sciences
Athene Square Grids, Electron Microscopy Sciences
Square Grids, Nickel, 3.05 mm, 300
Price
$155.01
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Highest quality square mesh transmission electron microscope grids.
  • Copper and nickel grids
  • 50, 150, 200, 300, and 400 mesh grids
  • Good handling characteristics
Individually inspected before being packed.
Spécifications
  • Ø:
    3.05 mm
  • Désignation:
    Square Grids
  • Matière:
    Nickel
  • Mesh:
    300