Athene Square Grids, Electron Microscopy Sciences
Athene Square Grids, Electron Microscopy Sciences
Square Grids, Copper, 3.05 mm, 150
  103302-118
Supplier #:  A150-CU
Athene Square Grids, Electron Microscopy Sciences
  103302-118
Athene Square Grids, Electron Microscopy Sciences
Athene Square Grids, Electron Microscopy Sciences
Square Grids, Copper, 3.05 mm, 150
$135.97
Each
Highest quality square mesh transmission electron microscope grids.
  • Copper and nickel grids
  • 50, 150, 200, 300, and 400 mesh grids
  • Good handling characteristics
Individually inspected before being packed.
  • Ø:
    3.05 mm
  • Désignation:
    Square Grids
  • Matière:
    Copper
  • Mesh:
    150