Chemical Containers, Electron Microscopy Sciences
Product Details & Documents
Relied on to transport dust samples from the moon, these injection-molded polyethylene containers are ideal for small quantities of liquids or solids.
- Made of injection-molded polyethylene
- Screw caps feature a leakproof inner seal and prevent leakage and spill
- Easy to clean
Spécifications
- Ø ext.:22 mm
- Capacité:5 ml
- Désignation:Container