Stainless Steel Baskets with Multiple Compartments, Electron Microscopy Sciences
Stainless Steel Baskets with Multiple Compartments, Electron Microscopy Sciences
4 Compartment Tissue Basket with Lid, 25 mm×23 mm
Catalog # 103302-066
Marketsource
Supplier #:  70194-29
Stainless Steel Baskets with Multiple Compartments, Electron Microscopy Sciences
Catalog # 103302-066
Marketsource
Stainless Steel Baskets with Multiple Compartments, Electron Microscopy Sciences
Stainless Steel Baskets with Multiple Compartments, Electron Microscopy Sciences
4 Compartment Tissue Basket with Lid, 25 mm×23 mm
Price
$163.25
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These stainless steel baskets with lids and multiple compartments are used for critical point drying.
  • Single or multiple compartments
These baskets are either 15 or 23 mm in diameter.
Specifications
  • Désignation:
    4 Compartment Tissue Basket with Lid
  • I.D.×H:
    25 mm×23 mm