3 D Stereo Glasses, Electron Microscopy Sciences
Product Details & Documents
White cardboard frames with temples for Red-Cyan or Red-Blue 3D anaglyph needs.
With red and cyan filters or Red and Blue filters, the glasses can be used for either red left or red right depending on which way you fold the temples.
Two different fold-scores allows to fit the glasses to smaller or larger viewer's heads.
Spécifications
- Désignation:3D Glasses, Red-Cyan