Chemical Containers, Electron Microscopy Sciences
Chemical Containers, Electron Microscopy Sciences
90 ml, Container, 52 mm
Catalog # 103301-994
Supplier #:  64299-43
Chemical Containers, Electron Microscopy Sciences
Catalog # 103301-994
Chemical Containers, Electron Microscopy Sciences
Chemical Containers, Electron Microscopy Sciences
90 ml, Container, 52 mm
Price
193,75 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Relied on to transport dust samples from the moon, these injection-molded polyethylene containers are ideal for small quantities of liquids or solids.
  • Made of injection-molded polyethylene
  • Screw caps feature a leakproof inner seal and prevent leakage and spill
  • Easy to clean
Specifications
  • Ø ext.:
    52 mm
  • Capacité:
    90 ml
  • Désignation:
    Container