Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences | Avantor
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Sampling Spoon, 17.8 cm (7"), 20 ml
Catalog # 103301-930
Fournisseur #:  72139
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Catalog # 103301-930
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Sampling Spoon, 17.8 cm (7"), 20 ml
Price
$43.63
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Retrieve small, specific volume samples of powders or liquids.
  • Wide handles
  • Unique flat bottom shape
  • Dual use as weighing boats
  • Non-sterile
  • Translucent
Long-handled spoons are ideal for acquiring samples from bottles and jars and helping to avoid contact with sample materials. A flat edge on the rim of each bowl allows fast leveling to collect and measure small amounts with accuracy.
Spécifications
  • Capacité:
    20 ml
  • Désignation:
    Sampling Spoon
  • Longueur:
    17.8 cm (7")