Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Sampling Spoon, 17.8 cm (7"), 5 ml
  103301-926
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
  103301-926
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Sampling Spoon, 17.8 cm (7"), 5 ml
$43.63
Each
Retrieve small, specific volume samples of powders or liquids.
  • Wide handles
  • Unique flat bottom shape
  • Dual use as weighing boats
  • Non-sterile
  • Translucent
Long-handled spoons are ideal for acquiring samples from bottles and jars and helping to avoid contact with sample materials. A flat edge on the rim of each bowl allows fast leveling to collect and measure small amounts with accuracy.
  • Capacité:
    5 ml
  • Désignation:
    Sampling Spoon
  • Longueur:
    17.8 cm (7")