EMS-Core Sampling Tools, Electron Microscopy Sciences
Product Details & Documents
EMS-Core is an all-new revolutionary multi-purpose sampling tool, designed with retractable cutting cannula for easy operation.
- Pen-like Actuator mechanically extends the cannula from the protective casing
- Self-contained Plunger effortlessly ejects the tissue sample out of the tip
- Polypropylene body, 304 stainless steel cutting canula (tip)
Efficient, safe, fast and precise, the EMS-Core consists of a razor-sharp stainless-steel cutting tip, designed to cut, retrieve and store cored samples from source materials such as skin, tissue, gels, paper, cloth, leaves, paint chips, films or other thin, soft substrates. The EMS-Core is well known and recommended as the special DNA sampling tool and is ideal for forensic and laboratory sampling purposes.
Specifications
- Désignation:Sampling Tool, 5 mm