Bacdown Handsoap, Decon Labs
Cleansing gel in poly bottle, 500 ml (16.9 oz.)
Supplier: Decon Labs
Fournisseur #: 7018
Jump to:
Product Details & Documents
Bacdown Handsoap combines soothing, coconut-based surfactants to provide a gentle handsoap that may be used repeatedly with minimal skin drying
- Soothing, gentle coconut-based surfactants
- Can be used repeatedly with minimal skin drying
- Pleasant almond fragrance
It is particularly recommended for applications where repeated hand scrubbing is mandated. Bacdown Handsoap also has a pleasant almond fragrance.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Capacité:500 ml (16.9 oz.)
- Désignation:Cleansing gel in poly bottle