Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Bacdown Handsoap, Decon Labs | Avantor

Bacdown Handsoap, Decon Labs

Cleansing gel in poly bottle, 1000 ml (34 oz.)
Catalog # CA89233-194
Supplier:   Decon Labs
Fournisseur #:  7001
Bacdown Handsoap, Decon Labs
Catalog # CA89233-194
Supplier:  Decon Labs
Bacdown Handsoap, Decon Labs
Bacdown Handsoap, Decon Labs

Cleansing gel in poly bottle, 1000 ml (34 oz.)

Price
$47.40
Log in pour la disponibilité
Each(1l) Case of 12 (1 l)
Product Details & Documents

Bacdown Handsoap combines soothing, coconut-based surfactants to provide a gentle handsoap that may be used repeatedly with minimal skin drying

  • Soothing, gentle coconut-based surfactants
  • Can be used repeatedly with minimal skin drying
  • Pleasant almond fragrance

It is particularly recommended for applications where repeated hand scrubbing is mandated. Bacdown Handsoap also has a pleasant almond fragrance.

Documents

Spécifications
  • Capacité:
    1000 ml (34 oz.)
  • Désignation:
    Cleansing gel in poly bottle