Dissolution Calibration Tools, Quality Lab Accessories
25 mm Digital Depth Gauge with Case, Multi-Instrument
75849-200
: QLA
: DDAPT-PLUS
QLA manufactures Calibrations tools that have been designed for use on most dissolution baths.
- Mechanical calibration parameters are easily verified using QLA's family of Calibration Tools
All are supplied with NIST traceable calibration certificates and user manuals. These calibration tools are serialized and come with a 1 year calibration certificate. Recalibration services are also offered by QLA.
: All calibration tools are serialized and come with a 1 year NIST traceable calibration certificate.
- Désignation:25 mm Digital Depth Gauge with Case, Multi-Instrument