Miscellaneous Dissolution Accessories, Quality Lab Accessories
Thermal Paper Rolls for Agilent, 2.25" wide (Pack/25)
  75848-982
 :   QLA
 :  TPP082-70XDS
Miscellaneous Dissolution Accessories, Quality Lab Accessories
  75848-982
 :  QLA
Miscellaneous Dissolution Accessories, Quality Lab Accessories
Miscellaneous Dissolution Accessories, Quality Lab Accessories
Thermal Paper Rolls for Agilent, 2.25" wide (Pack/25)
$307.92
Pack of 25
Everclear Water Bath Preservative is a crystalline formula offered in premeasured vials for use with dissolution water baths.Use with DEIONIZED water
The following products are compatible with Agilent/VanKel equipment: APP7 accessories, Rotating Bottle accessories, Auto-sampler accessories and Paper Rolls.
PTFE Suspension cups can be used with most dissolution baths.
A single dose will protect a bath for up to 8 weeks.
  • Désignation:
    Thermal Paper Rolls for Agilent, 2.25" wide (Pack/25)