Disintegration Testing, Quality Lab Accessories
Glass Replacement Tubes for 3 Tube Disintegration Assembly, 38 mm
  75848-894
 :   QLA
 :  DISTUB-03
Disintegration Testing, Quality Lab Accessories
  75848-894
 :  QLA
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Glass Replacement Tubes for 3 Tube Disintegration Assembly, 38 mm
$311.62
Pack of 3
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
  • Désignation:
    Glass Replacement Tubes for 3 Tube Disintegration Assembly, 38 mm