Disintegration Testing, Quality Lab Accessories | Avantor
Disintegration Testing, Quality Lab Accessories
10 Mesh SS Screens for 6 Tube Disintegration Assembly
Catalog # 75848-892
Supplier:   QLA
Fournisseur #:  DISSCR-VK10
Disintegration Testing, Quality Lab Accessories
Catalog # 75848-892
Supplier:  QLA
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
10 Mesh SS Screens for 6 Tube Disintegration Assembly
Price
$90.15
Log in pour la disponibilité
Pack of 6
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Spécifications
  • Désignation:
    10 Mesh SS Screens for 6 Tube Disintegration Assembly