Disintegration Testing, Quality Lab Accessories
10 Mesh SS Screens for 6 Tube Disintegration Assembly
Catalog # 75848-892
Supplier: QLA
Fournisseur #: DISSCR-VK10
Jump to:
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
- Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Spécifications
- Désignation:10 Mesh SS Screens for 6 Tube Disintegration Assembly
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.