Disintegration Testing, Quality Lab Accessories
USP Fluted Plastic Disks for 3 Tube Disintegration Assembly
Catalog # 75848-888
Supplier: QLA
Fournisseur #: DISDISK-03
Jump to:
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
- Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Spécifications
- Désignation:USP Fluted Plastic Disks for 3 Tube Disintegration Assembly