Disintegration Testing, Quality Lab Accessories | Avantor
Disintegration Testing, Quality Lab Accessories
Disintegration Basket Assembly with 3 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
Catalog # 75848-886
Supplier:   QLA
Fournisseur #:  DISASY-03
Disintegration Testing, Quality Lab Accessories
Catalog # 75848-886
Supplier:  QLA
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Disintegration Basket Assembly with 3 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
Price
$2,042.96
Log in pour la disponibilité
Each
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Spécifications
  • Désignation:
    Disintegration Basket Assembly with 3 Glass Tubes and 10 Mesh SS Screens for VanKel Testers