Diesse Mini-Cube ESR Analyzer, Streck
Diesse ESR printer
Catalog # 75845-764
Supplier:   Streck Laboratories
 :  240402
Diesse Mini-Cube ESR Analyzer, Streck
Catalog # 75845-764
Diesse Mini-Cube ESR Analyzer, Streck
Diesse Mini-Cube ESR Analyzer, Streck
Diesse ESR printer
Price
$1,025.37
Log in pour la disponibilité
Each
Product Details & Documents
An automated instrument for erythrocyte sedimentation rate testing in EDTA tubes.
  • Produces ESR Results directly from EDTA tubes without consuming patient samples
  • Compatible with standard 13×75 mm EDTA Tubes (2.0 to 4.0 ml sample volume) with HemogardTM or conventional stoppers, and 500 µl EDTA microtainer tubes
  • Random access
  • Results in 20 minutes
  • >95% correlation to modified westergren method
 : A two-level hematology control used to monitor erythrocyte sedimentation rates on automated methods that use EDTA tubes
Manufactured from human red blood cells
Contains barcodes for automatic QC file archive
Alerts the technologists to possible problems that may affect the accuracy of patient results
12-months closed-vial stability
7-day open-vial stability at room temp
Printer is Bluetooth enabled and automatically prints results
Spécifications
  • Désignation:
    Diesse ESR printer