Diesse Mini-Cube ESR Analyzer, Streck | Avantor
Diesse Mini-Cube ESR Analyzer, Streck
Diesse printer paper
Catalog # 75845-760
Supplier:   Streck Laboratories
Fournisseur #:  240414
Diesse Mini-Cube ESR Analyzer, Streck
Catalog # 75845-760
Diesse Mini-Cube ESR Analyzer, Streck
Diesse Mini-Cube ESR Analyzer, Streck
Diesse printer paper
Price
$91.32
Log in pour la disponibilité
Pack of 5
La quantité minimale de commande est de 1
Product Details & Documents
An automated instrument for erythrocyte sedimentation rate testing in EDTA tubes.
  • Produces ESR Results directly from EDTA tubes without consuming patient samples
  • Compatible with standard 13×75 mm EDTA Tubes (2.0 to 4.0 ml sample volume) with HemogardTM or conventional stoppers, and 500 µl EDTA microtainer tubes
  • Random access
  • Results in 20 minutes
  • >95% correlation to modified westergren method
Informations sur les accessoires: A two-level hematology control used to monitor erythrocyte sedimentation rates on automated methods that use EDTA tubes
Manufactured from human red blood cells
Contains barcodes for automatic QC file archive
Alerts the technologists to possible problems that may affect the accuracy of patient results
12-months closed-vial stability
7-day open-vial stability at room temp
Printer is Bluetooth enabled and automatically prints results
Spécifications
  • Désignation:
    Diesse printer paper