BOD Bottles
300 ml
  470223-794
 :  BOD300
BOD Bottles
  470223-794
BOD Bottles
BOD Bottles
300 ml
32,70 $
Each Pack of 24
For biochemical oxygen demand testing.
  • Glass bottles
  • Bottles are unnumbered
  • Feature penny head stopper
BOD Bottles are designed for incubating diluted samples of clean water, waste water, effluent water, and polluted water during the five-day biochemical oxygen demand (BOD) test.
  • Capacité:
    300 ml