BOD Bottles
60 ml
Catalog # 470223-792
Supplier: United Scientific Supplies
Fournisseur #: BOD060
Jump to:
Product Details & Documents
For biochemical oxygen demand testing.
- Glass bottles
- Bottles are unnumbered
- Feature penny head stopper
BOD Bottles are designed for incubating diluted samples of clean water, waste water, effluent water, and polluted water during the five-day biochemical oxygen demand (BOD) test.
Spécifications
- Capacité:60 ml