BOD Bottles | Avantor
BOD Bottles
60 ml
Catalog # 470223-792
Fournisseur #:  BOD060
BOD Bottles
Catalog # 470223-792
BOD Bottles
BOD Bottles
60 ml
Price
$25.75
Log in pour la disponibilité
Each Pack of 36
Product Details & Documents
For biochemical oxygen demand testing.
  • Glass bottles
  • Bottles are unnumbered
  • Feature penny head stopper
BOD Bottles are designed for incubating diluted samples of clean water, waste water, effluent water, and polluted water during the five-day biochemical oxygen demand (BOD) test.
Spécifications
  • Capacité:
    60 ml