Student Thermometer
−20…+110 °C (0...230 °F), 30 cm (11¾"), Partial
  470223-768
 :  THPCF1
Student Thermometer
  470223-768
Student Thermometer
Student Thermometer
−20…+110 °C (0...230 °F), 30 cm (11¾"), Partial
$12.80
Each
No-roll triangle shape.
  • Partial immersion
  • White backing
  • Features re-usable plastic storage tube
This student grade thermometer is filled with kerosene with red dye.
  • Longueur:
    30 cm (11¾")
  • Range:
    −20…+110 °C (0...230 °F)
  • Immersion:
    Partial