Student Thermometer
−20…+110 °C (0...230 °F), 30 cm (11¾"), Partial
470223-768
: THPCF1
No-roll triangle shape.
- Partial immersion
- White backing
- Features re-usable plastic storage tube
This student grade thermometer is filled with kerosene with red dye.
- Longueur:30 cm (11¾")
- Range:−20…+110 °C (0...230 °F)
- Immersion:Partial