CELLROLL Roller Bottle System, Argos Technologies | Avantor
CELLROLL Roller Bottle System, Argos Technologies
CELLROLL Interface Cable
Catalog # 75834-458
CELLROLL Roller Bottle System, Argos Technologies
Catalog # 75834-458
Supplier:  
CELLROLL Roller Bottle System, Argos Technologies
CELLROLL Roller Bottle System, Argos Technologies
CELLROLL Interface Cable
Discontinued
Product Details & Documents
CELLROLL a reliable cell culture system.
  • Programmable control unit
  • Rotation speeds of either 1-2 rpm or 2-6 rpm
  • Equipped with an alarm system
The modular design of the CELLROLL allows the user to easily scale-up production depending on the needs: one motor unit can control between one and eight bottle racks, each rack accommodating two standard roller bottles. The moisture-resistant motor unit is available with rotation speeds of either 1-2 rpm or 2-6 rpm and is steered by a programmable control unit located outside of the incubator. In case of power failure or accidental disconnection of control and motor unit an alarm system is activated. The corrosion-free and sturdy construction makes the CELLROLL a reliable cell culture system that will keep your cells always "on the move.
Certifications: CE certified.
Ordering information: Includes drive unit, two decks (for 4 roller bottles), a control unit, a power supply and a control cable.
Spécifications
  • Désignation:
    CELLROLL Interface Cable
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.