Lambda PFG Ladder, New England Biolabs
Lambda PFG Ladder, 50 gel lanes
103218-940
: N0341S
Lambda PFG Ladder consists of one GelSyringe™ dispenser, sufficient for 50 gel lanes
- Suitable for pulsed field gel electrophoresis
- Size range: 48.5 to 1018 kb
- Includes one GelSyringe dispenser
- Suitable for 50 gel lanes
Successively larger concatemers of lambda DNA (cI857 ind 1 Sam7) are embedded in 1% LMP agarose. This product is designed to be used as size markers for pulsed-field gel electrophoresis (PFG).
- Désignation:Lambda PFG Ladder
- Taille:50 gel lanes