Lambda PFG Ladder, New England Biolabs
Lambda PFG Ladder, 50 gel lanes
  103218-940
 :  N0341S
Lambda PFG Ladder, New England Biolabs
  103218-940
Lambda PFG Ladder, New England Biolabs
Lambda PFG Ladder, New England Biolabs
Lambda PFG Ladder, 50 gel lanes
$300.20
Each
Lambda PFG Ladder consists of one GelSyringe™ dispenser, sufficient for 50 gel lanes
  • Suitable for pulsed field gel electrophoresis
  • Size range: 48.5 to 1018 kb
  • Includes one GelSyringe dispenser
  • Suitable for 50 gel lanes
Successively larger concatemers of lambda DNA (cI857 ind 1 Sam7) are embedded in 1% LMP agarose. This product is designed to be used as size markers for pulsed-field gel electrophoresis (PFG).
  • Désignation:
    Lambda PFG Ladder
  • Taille:
    50 gel lanes