Trio.Bas™ MINI Kits
Trio.Bas™ Mini air sampler, with bluetooth, 200 L/min, Petri plate
  75804-684
 :  BAS163
Trio.Bas™ MINI Kits
  75804-684
Trio.Bas™ MINI Kits
Trio.Bas™ MINI Kits
Trio.Bas™ Mini air sampler, with bluetooth, 200 L/min, Petri plate
$12,690.00
Each
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
  • Aspirating head with easy manipulation bayonet closure
  • Manual or automatic operations
  • Ergonomic and balanced design to facilitate handling with ease
All of the air samplers are lightweight, with an antibacterial techno-polymer shockproof body. They have a long lasting battery and are IP65 protected from dust and water.
Per USP 797, 'Impaction is the preferred method of active air sampling. Use of settling plates for qualitative air sampling cannot be relied upon and shall not be used solely to determine the quality of air in the controlled environment.'
 : ISO14698, USP 797, USP 116, EU GPL-GMP, ISO 17025.
 : Additional accessory components, including data transfer options, are available separately.
 : Trio.Bas™ Mini kit contains:
Trio.Bas™ Mono air sampler with Bluetooth data transfer, includes calibration certificate
100/240 V battery charger with cable and adapter
Stainless steel aspirating head, autoclavable
Light carrying case to protect sampler in transit
  • Débit:
    200 L/min
  • Désignation:
    Trio.Bas™ Mini air sampler, with bluetooth
  • Head Type:
    Petri plate
Services
  •  
    Description
  • 76308-450
    Calibration Service for MINI and MONO air sampler
    Each - $2,220.00