$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
FUNNEL CELL DBL DISP OFFSET F/CLIP SYS
75802-106
:
HETTICH INSTRUMENTS
FUNNEL CELL DBL DISP OFFSET F/CLIP SYS
75802-106
:
HETTICH INSTRUMENTS
:
7215
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International